missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosyladenine, 99%, Thermo Scientific Chemicals

Catalog Number 15456709
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
5536-17-4
C10H13N5O4
267.25
MFCD00065471,MFCD00065471,MFCD00475667,MFCD00005752,MFCD00065471
OIRDTQYFTABQOQ-UHFFFAOYNA-N
3s,4r,5r-2-6-aminopurin-9-yl-5-hydroxymethyl oxolane-3,4-diol, prestwick_983, 9-lyxofuranosyladenine
6420052
(3S,4R,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
NC1=C2N=CN(C3OC(CO)C(O)C3O)C2=NC=N1
This item is not returnable. View return policy
5536-17-4
C10H13N5O4
267.25
MFCD00065471,MFCD00065471,MFCD00475667,MFCD00005752,MFCD00065471
OIRDTQYFTABQOQ-UHFFFAOYNA-N
3s,4r,5r-2-6-aminopurin-9-yl-5-hydroxymethyl oxolane-3,4-diol, prestwick_983, 9-lyxofuranosyladenine
6420052
(3S,4R,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
NC1=C2N=CN(C3OC(CO)C(O)C3O)C2=NC=N1

Vidarabine is a neurotransmitter that acts as the preferred endogenous agonist at all adenosine receptor subtypes. Vidarabine is an inhibitor of A cyclase and DNA Polymerase. Used to study the roles of AMP-activated protein kinase (AMPK) in cell signaling. Cell-permeable adenylate cyclase inhibitor; in detergent-dispersed rat brain preparation, IC50 = 30 ?M. Clinically significant antiviral agent, especially against herpes simplex (HSV),1 by inhibition of DNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vidarabine is a neurotransmitter that acts as the preferred endogenous agonist at all adenosine receptor subtypes. Vidarabine is an inhibitor of A cyclase and DNA Polymerase. Used to study the roles of AMP-activated protein kinase (AMPK) in cell signaling. Cell-permeable adenylate cyclase inhibitor; in detergent-dispersed rat brain preparation, IC50 = 30 μM. Clinically significant antiviral agent, especially against herpes simplex (HSV),1 by inhibition of DNA polymerase.

Solubility
Soluble in DMF (10 mg/ml), 0.5 M HCl (50 mg/ml), DMSO (53 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), and water (3 mg/ml at 25°C).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Beilstein 624881
Merck Index 14,9976
Solubility Information Soluble in DMF (10mg/ml),0.5 M HCl (50mg/ml),DMSO (53mg/ml at 25°C),ethanol (<1mg/ml at 25°C),and water (3mg/ml at 25°C).
Formula Weight 267.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 9-beta-D-Arabinofuranosyladenine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'