missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-6-chloroimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals

Catalog Number 15417467
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
957187-27-8
C7H4BrClN2
231.48
MFCD12828062
LMPPBTNPACXNDI-UHFFFAOYSA-N
8-bromo-6-chloroimidazo 1,2-a pyridine, 8-bromo-6-chloro-imidazo 1,2-a pyridine, 6-chloro-8-bromoimidazo 1,2-a pyridine
26370038
8-bromo-6-chloroimidazo[1,2-a]pyridine
ClC1=CN2C=CN=C2C(Br)=C1
This item is not returnable. View return policy
957187-27-8
C7H4BrClN2
231.48
MFCD12828062
LMPPBTNPACXNDI-UHFFFAOYSA-N
8-bromo-6-chloroimidazo 1,2-a pyridine, 8-bromo-6-chloro-imidazo 1,2-a pyridine, 6-chloro-8-bromoimidazo 1,2-a pyridine
26370038
8-bromo-6-chloroimidazo[1,2-a]pyridine
ClC1=CN2C=CN=C2C(Br)=C1

8-Bromo-6-chloroimidazo[1,2-a]pyridine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Bromo-6-chloroimidazo[1,2-a]pyridine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 125°C to 128°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 231.48
Percent Purity 95%
Chemical Name or Material 8-Bromo-6-chloroimidazo[1,2-a]pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'