Learn More
6-Methyl-5-nitroquinoline, 97%, Thermo Scientific™
Catalog Number 11360234
Click to view available options
Quantity:
1 g
5 g
23141-61-9
C10H8N2O2
188.186
MFCD00024021
CENBTULRDDPOGR-UHFFFAOYSA-N
5-nitro-6-methylquinoline, quinoline, 6-methyl-5-nitro, quinoline,6-methyl-5-nitro, acmc-1cuis, maybridge1_002137, 5-nitro-6-met hylquinoline, 5-nitro-6-methyl quinoline, 6-methyl-5-nitroquinoline
90009
6-methyl-5-nitroquinoline
CC1=C(C2=C(C=C1)N=CC=C2)[N+](=O)[O-]
23141-61-9
C10H8N2O2
188.186
MFCD00024021
CENBTULRDDPOGR-UHFFFAOYSA-N
5-nitro-6-methylquinoline, quinoline, 6-methyl-5-nitro, quinoline,6-methyl-5-nitro, acmc-1cuis, maybridge1_002137, 5-nitro-6-met hylquinoline, 5-nitro-6-methyl quinoline, 6-methyl-5-nitroquinoline
90009
6-methyl-5-nitroquinoline
CC1=C(C2=C(C=C1)N=CC=C2)[N+](=O)[O-]
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Melting Point 115°C to 118°C
Quantity 1 g
Beilstein 154688
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 6-Methyl-5-nitroquinoline