Learn More
Greiner Bio-One 50 mL Sterile Serological Pipets
GREENER_CHOICE
Product Code. 10561804
Change view
Click to view available options
Packaging Type:
Individual Plastic Wrap
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Type unitSize
10561804 Individual Plastic Wrap Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10561804 Supplier Greiner Bio-One Supplier No. 768160
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Drip-free pipetting design with high optical clarity and maximum accuracy.
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
TRUSTED_SUSTAINABILITY
Specifications
Aspirator Plug Yes
Autoclavable Not Autoclavable
Description Shape: Standard
Material Polystyrene
Sterility Sterile
For Use With (Application) Sterile Liquid Handling
Graduated Yes
Graduations 1/2
Includes Synthetic Fibrous Filter Material
Negative Graduations Yes
Quantity 100/Cs.
Packaging Type Individual Plastic Wrap
Volume (Metric) 50 mL
Type Serological Pipet
Disposable Single-use
Color Code Purple