Learn More
Brady™ IP Printer Enabled 4500 Series Thermal Transfer Printer Ribbons
Offers environmental resistance against smearing and chemicals
Brand:  Brady™ 066189
Additional Details : Weight : 0.27670kg
 View more versions of this product
Product Code. 11593025
  • 232.00€ / 1 box
Estimated Shipment: 09-07-2024
Description
  • Sharp-edge print quality to maximize bar code readability
  • Includes spindle
  • Compatible with IP300, IP600 printers
  • Core inside diameter: 25.40mm (1 in.), ink side out
Specifications
Specifications
Thermal Transfer Printer Ribbon
IP300 Thermal Transfer Printer, IP600 Thermal Transfer Printer
984 in.
300 m
25.4 mm I.D.
Silver 4500 Series Thermal Transfer Printer Ribbon - Brady IP Printer Enabled
Ink side out
1 Ea.
Silver
Wax
3.27 in.
8.3 cm
300 m x 8.3 cm (984 ft. x 3.27 in.)
IP™ Enabled
RoHS 2002/95/EC