Learn More
STEMCELL Technologies 3T3-J2 Irradiated Feeder Cells
Irradiated 3T3-J2 mouse fibroblast, frozen
Brand: STEMCELL Technologies 100-0353
Additional Details : Weight : 0.50000kg
Description
- 3T3-J2 Irradiated Feeder Cells are optimized to support epithelial cell expansion in Conditional Reprogramming (CR) Medium (Catalog #100-0352; Liu X et al. 2012; Liu X et al. 2017; Suprynowicz FA et al.) The cells are mitotically inactivated by radiation, but maintain metabolic activity
- 3T3-J2 Irradiated Feeder Cells are cryopreserved and can be used directly for seeding epithelial cells without plating in advance
- Each vial contains 2 - 3 x 10^6 cells and can be used to seed up to three T-25 cm2 flasks or one T-75 cm2 flask
- 3T3-J2 Irradiated Feeder Cells are intended for Research Use Only
Specifications
Cell Culture, Differentiation, Expansion | |
0.5 mL |