Learn More
3M™ 6000 Series Half Face Mask Respirator
Half-face mask respirators 6000 Series are economical, low-maintenance, and extremely lightweight.
Brand: 3M™ 7000146849
Additional Details : Weight : 0.16200kg
Description
Combines features of both reusable and disposable respirators to provide effective protection. 3M™ Half Face Mask Respirator 6000 Series are economical, low-maintenance, simple-to-handle, and extremely lightweight.
- Reusable half masks are economical, low-maintenance, simple-to-handle, and extremely lightweight
- Used with 3M™ Supplied Air Systems
- Bayonet Connection System allows connection to a broad range of twin lightweight filters to protect against gases, vapors, and particulates
- Twin filter design provides lower breathing resistance
- Probed reusable respirator assembly 6000 Series used for quantitative fit testing (17-987-3A & -3B)
Compliance
NIOSH approved under 42 CFR 84 standard for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters; in Canada, CSA standard Z94.4 and applicable local government requirements must be met for approval.
Specifications
EN 140:1998 | |
Bayonet | |
Thermoplastic Elastomer | |
Large | |
Cartridge 6000 Series, Filter 2000, 2200, 7000 or 5000 Series, or Dual Airline System | |
6300 |
NIOSH 42 CFR Part 84 | |
Gray | |
Half-Mask Respirator | |
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding | |
Reclosable storage bag | |
4-point |