Learn More
Bochem™ 3-Finger Laboral Retort Clamp
Product Code. 10228980
Change view
Click to view available options
Diameter (Metric):
12 to 100 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10228980 12 to 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10228980 Supplier Bochem™ Supplier No. B08256108
In Stock
Add to basket
Edge
Add to basket
Coated with soft PVC, the three fingers of this independently adjustable retort clamp are well suited for holding delicate objects. Bochem™ 3-Finger Laboral Retort Clamp has a base construction of Laboral aluminum alloy.
  • Laboral aluminum alloy construction.
  • PVC-coated fingers.
  • Span width is independently adjustable with 2 regulating screws.
Specifications
Quantity 1 Ea.
Type Stand Clamp
Material Laboral, PVC
Diameter (Metric) 12 to 100 mm
Diameter (Metric) Outer Tubing 12 mm
Length (Metric) 140 mm
Length (Metric) Arm 140 mm
Size (Metric) Grip 12 to 100 mm
Jaw Type 3-Prong
Diameter (Metric) Inside 12 to 100 mm