Learn More
3-Butyn-2-one, 96%, ACROS Organics™
Brand: Acros Organics 330440500
Additional Details : CAS : 1423-60-5 Weight : 0.10000kg Transport : UN number : 1992 Chem class : 3 Pack group : II
Packaging | Glass Bottle |
---|---|
Quantity | 50g |
Chemical Identifiers
1423-60-5 | |
68.06 | |
3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch | |
CHEBI:48060 | |
CC(=O)C#C |
C4H4O | |
XRGPFNGLRSIPSA-UHFFFAOYSA-N | |
15018 | |
but-3-yn-2-one |
Specifications
3-Butyn-2-one | |
−1°C | |
Glass Bottle | |
0.87 | |
Undesignated | |
1423-60-5 | |
C4H4O | |
03,38 | |
XRGPFNGLRSIPSA-UHFFFAOYSA-N | |
but-3-yn-2-one | |
15018 | |
68.06 | |
96% |
0.8700g/mL | |
Authentic | |
1.4045 to 1.4095 | |
85.0°C | |
50g | |
95% min. (GC) | |
HC≡CCOCH3 | |
3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch | |
CC(=O)C#C | |
68.06 | |
CHEBI:48060 | |
Liquid |
Safety and Handling
- 3-Butyn-2-one
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H300-Fatal if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.