Learn More
Thermo Scientific™ 250μL PTFE Luer-Lok, Gas-Tight Syringe
Thermo Scientific Luer Syringes offer easy, accurate and reproducible injections for a variety of volumes.
Brand:  Thermo Scientific™ 365I7658
Additional Details : Weight : 0.10000kg
Product Code. 15204593
  • 64.17€ / Each
Estimated Shipment: 03-06-2024
Specifications
Gas Tight GC Autosampler Syringe, PTFE Luer-Lok
250 μL
Gas Chromatography Systems