Learn More
Fisherbrand™ 22mm PE, Cap, Transparent, Center hole
Polyethylene caps are chemically inert and suitable for most chromatography applications.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16338387
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • 22mm PE Cap
  • Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
Specifications
Specifications
Transparent
Polyethylene
100
ND22
22 mm
9.1 mm
Snap Cap