Learn More
HUBER Insulated Stainless Steel Bath 208B
Insulated bath
Brand:  HUBER 6683
Additional Details : Weight : 8.10000kg
Product Code. 10772664
  • 749.42€ / Each
Estimated Shipment: 16-07-2024
Description
Insulated stainless steel bath 208B, 8.5L
Specifications
Specifications
8.5 L
200°C
208 B