Learn More
Linker Industrietechnik™ 200 Dia. x 45mm Height Plastic Polyoxymethylene Frame Test Sieves
Brand: Linker Industrietechnik™ 552.050.881-0015
Additional Details : Weight : 0.38000kg
Specifications
Nylon | |
1Each | |
Test Sieve | |
Polyoxymethylene |
200 mm | |
15μm | |
DIN 4197 | |
45mm |