Learn More
2-Methyl-2-pentenoic acid, 99%, Thermo Scientific Chemicals
Catalog Number p-7023236
Click to view available options
Quantity:
10 g
50 g
250 g
3142-72-1
C6H10O2
114.144
MFCD00002655
JJYWRQLLQAKNAD-SNAWJCMRSA-N
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
5365909
(E)-2-methylpent-2-enoic acid
CCC=C(C)C(=O)O
3142-72-1
C6H10O2
114.144
MFCD00002655
JJYWRQLLQAKNAD-SNAWJCMRSA-N
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
5365909
(E)-2-methylpent-2-enoic acid
CCC=C(C)C(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 0.978
Boiling Point 120°C (20 mmHg)
Flash Point 107°C (225°F)
Odor Characteristic
Quantity 10 g
UN Number UN3265
Formula Weight 114.15
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-pentenoic acid