Learn More
2-Chloropyridine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
2-Chloropyridine-5-carboxaldehyde is used as pharmaceutical intermediates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloropyridine-5-carboxaldehyde is used as pharmaceutical intermediates
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
2-Chloropyridine-5-carboxaldehyde is used as pharmaceutical intermediates
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | 77°C to 81°C |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 141.56 |
Percent Purity | 96% |
Chemical Name or Material | 2-Chloropyridine-5-carboxaldehyde |