Learn More
2,3-Difluorotoluene, 98%, Thermo Scientific™
Catalog Number 15595246
Click to view available options
Quantity:
1 g
5 g
25 g
3828-49-7
C7H6F2
128.12
MFCD01631506
ZNEHIDGAPGVZSA-UHFFFAOYSA-N
2,3-difluorotoluene, 1,2-difluoro-3-methyl-benzene, benzene, 1,2-difluoro-3-methyl, 2,3-difluoro methyl benzene, pubchem1610, 2,3-difluoro toluene, acmc-209iz4, 2.3-difluoro methyl benzene, ksc493c6t
2774228
1,2-difluoro-3-methylbenzene
CC1=CC=CC(F)=C1F
3828-49-7
C7H6F2
128.12
MFCD01631506
ZNEHIDGAPGVZSA-UHFFFAOYSA-N
2,3-difluorotoluene, 1,2-difluoro-3-methyl-benzene, benzene, 1,2-difluoro-3-methyl, 2,3-difluoro methyl benzene, pubchem1610, 2,3-difluoro toluene, acmc-209iz4, 2.3-difluoro methyl benzene, ksc493c6t
2774228
1,2-difluoro-3-methylbenzene
CC1=CC=CC(F)=C1F
Density 1.0960g/mL
Boiling Point 119.0°C to 121.0°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.096
Formula Weight 128.12
Percent Purity 98%
Chemical Name or Material 2,3-Difluorotoluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Difluorotoluene, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.