missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number 11339805
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
226396-32-3
C6H4BF3O2
175.901
MFCD01863168
CLGIPVVEERQWSQ-UHFFFAOYSA-N
2,3,4-trifluorophenyl boronic acid, 2,3,4-trifluorobenzeneboronic acid, 2,3,4-trifluorphenylboronic acid, 4,5,6-trifluorophenylboronic acid, boronic acid, 2,3,4-trifluorophenyl, pubchem1887, acmc-1ckzl, trifluorophenylboronic acid, ksc201q5d, jalor-chem i01-8460
2783280
(2,3,4-trifluorophenyl)boronic acid
B(C1=C(C(=C(C=C1)F)F)F)(O)O
This item is not returnable. View return policy
226396-32-3
C6H4BF3O2
175.901
MFCD01863168
CLGIPVVEERQWSQ-UHFFFAOYSA-N
2,3,4-trifluorophenyl boronic acid, 2,3,4-trifluorobenzeneboronic acid, 2,3,4-trifluorphenylboronic acid, 4,5,6-trifluorophenylboronic acid, boronic acid, 2,3,4-trifluorophenyl, pubchem1887, acmc-1ckzl, trifluorophenylboronic acid, ksc201q5d, jalor-chem i01-8460
2783280
(2,3,4-trifluorophenyl)boronic acid
B(C1=C(C(=C(C=C1)F)F)F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 202°C to 204°C
Quantity 250 mg
Formula Weight 175.9
Percent Purity 98%
Chemical Name or Material 2,3,4-Trifluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'