missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10666272
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
540-84-1
C8H18
114.232
8943
NHTMVDHEPJAVLT-UHFFFAOYSA-N
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
10907
CHEBI:62805
2,2,4-trimethylpentane
CC(C)CC(C)(C)C
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
540-84-1
C8H18
114.232
8943
NHTMVDHEPJAVLT-UHFFFAOYSA-N
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
10907
CHEBI:62805
2,2,4-trimethylpentane
CC(C)CC(C)(C)C
Melting Point -107°C
Color Colorless
Boiling Point 98°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 51mbar at 20°C
Viscosity 0.51 mPaS at 22°C
Formula Weight 114.23g/mol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'