Learn More
Abnova™ 19p13.3/19q13.13 FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0159.200uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16043255
  • 2430.00€ / 200µL
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Specifications
Store at 4°C in the dark.
Tube
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Liquid
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
200 μL