Learn More
Bochem™ 18/10 Stainless Steel Laboratory Spoon
Product Code. 10176360
Click to view available options
Length (Metric):
150 mm
170 mm
200 mm
Dimensions (L x W):
20 x 10 mm
30 x 15 mm
30 x 26 mm
40 x 10 mm
Unit Size:
Each
Product Code. 10176360
Brand: Bochem™ 3385
Edge
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Laboratory Spoon serve a wide variety of functions in the lab.
  • Lab spoon
  • Manufactured from 18/10 stainless steel
Specifications
Quantity 1 Ea.
Product Type Laboratory Spoon
Dimensions (L x W) 20 x 10 mm
Material Stainless Steel (18/10)
Length (Metric) 150 mm