Learn More
Bochem™ 18/10 Stainless Steel Forceps, Blunt Tip
Product Code. 10169920
Change view
Click to view available options
Length (Metric):
105 mm
115 mm
130 mm
145 mm
160 mm
200 mm
250 mm
300 mm
Unit Size:
Each
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10169920 130 mm Each
10579661 105 mm Each
10455641 115 mm Each
10393551 145 mm Each
10663341 160 mm Each
10768721 200 mm Each
11845121 200 mm Pack of 10
10758531 250 mm Each
10383601 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10169920 Supplier Bochem™ Supplier No. 1002
  • ONLINE EXCLUSIVE PRICE
    4.80€
    3.79€ / Pack of 1
    Save 1.01€
    21% Off
In Stock
Add to basket
Edge
Add to basket
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Forceps serve a wide variety of functions in the lab.
  • Stainless Steel
  • Various lengths
Specifications
Product Type Forceps
Length (Metric) 130 mm
Material Stainless Steel (18/10)
Quantity 1 Pack
Tip Style Blunt