Learn More
Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim
Evaporate materials in the laboratory wit the Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim.
Brand: Bochem™ 8399
Additional Details : Weight : 0.10000kg
Description
- 18/10 stainless steel
- Substantial rim size
- Rectangular shape
Specifications
Flat | |
Rectangular | |
500 mm |
Stainless Steel | |
Evaporating Dish | |
100 mm |