Learn More
Bochem™ 18/10 Stainless Steel Chemical Spoon
Product Code. 10485031
Change view
Click to view available options
Length (Metric):
150 mm
180 mm
210 mm
250 mm
300 mm
Length (Metric) Blade:
32 mm
40 mm
45 mm
48 mm
Diameter (Metric) Outer:
1 mm
2 mm
5 mm
Unit Size:
Each
Pack of 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Length (Metric) Blade Diameter (Metric) Outer unitSize
10485031 180 mm 32 mm 1 mm Each
10538881 150 mm 32 mm 5 mm Each
11865071 150 mm 32 mm 5 mm Pack of 10
10262091 150 mm 32 mm 1 mm Each
10343981 210 mm 32 mm 1 mm Each
10373741 180 mm 40 mm 5 mm Each
11855071 210 mm 40 mm 5 mm Pack of 10
10169960 250 mm 45 mm 2 mm Each
10300681 250 mm 48 mm 5 mm Each
10613341 300 mm 48 mm 5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10485031 Supplier Bochem™ Supplier No. 3422
In Stock
Add to basket
Edge
Add to basket
Stainless steel spoon for scooping chemicals.
  • Chemical spoon
  • With spatula handle
Specifications
Quantity 1 Ea.
Product Type Chemical Scoop
Length (Metric) Blade 32 mm
Width (Metric) 22 mm
Diameter (Metric) Outer 1 mm
Material Stainless Steel (18/10)
Length (Metric) 180 mm