Learn More
Bochem™ 18/10 Stainless Ladle Scoops with Flat Handle
Brand: Bochem™ 8790
Additional Details : Weight : 0.50000kg
Specifica
Ladle Scoop | |
Flat | |
300 mm |
Stainless Steel (18/10) | |
65 mL |