Learn More
Hamilton™ 1700 Series Removable Needle Syringes
Ideal for dispensing both liquids and gases.
116.00€ - 1615.00€
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
11507804
|
Hamilton™
80014 |
32 G |
116.00€
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11528024
|
Hamilton™
DS80065 |
22s G |
1615.00€
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
- Precision-machined PTFE plunger tip which creates a leak-free seal
- Tip wipes the interior of the syringe barrel free of sample
- Fluid path is chemically inert as possible
Spécification
10 μL |