Learn More
Helios Italquartz 1234 DIN Standard Crucibles, Transparent
1234 DIN standard crucibles constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand: Helios Italquartz 63P00515
Additional Details : Weight : 0.20000kg
Description
- Transparent
- Conform to 1234 DIN Standard
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Transparent | |
Quartz | |
60 mm | |
1234 DIN standard crucible |
Model CR 55 | |
90 cc | |
48 mm |