Learn More
Helios Italquartz 1234 DIN Standard Covers with Ring Handle, Transparent
1234 DIN standard covers with ring handle constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 64P00607
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15848094
  • 155.00€ / Each
Estimated Shipment: 01-08-2024
Description
  • Transparent
  • Include ring handle
  • Conform to 1234 DIN standard
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Transparent
Quartz
1234 DIN Standard Covers with Ring Handle
CR 47-56 Crucible