Learn More
Helios Italquartz 1234 DIN Standard Covers with Ring Handle, Transparent
1234 DIN standard covers with ring handle constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand: Helios Italquartz 64P00606
Additional Details : Weight : 0.20000kg
Description
- Transparent
- Include ring handle
- Conform to 1234 DIN standard
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Transparent | |
Quartz | |
1234 DIN Standard Covers with Ring Handle | |
CR 46-55-66 Crucible |