Learn More
Sartorius™ 11406 Series Gridded Sterile Cellulose Nitrate (CN) Membrane Filters
11406 Series Gridded Sterile Cellulose Nitrate (CN) Membrane Filters have a very uniform pore structure and high non-specific adsorption. Single-packed or dispenser-ready, these individually wrapped filters are ideal for colony counting, particle testing, microscopy, and sterility testing.
144.00€ - 217.56€
Specifications
Color | White |
---|---|
Disposable | Single-use |
Filter Type | Membrane Filter |
Format | Disc |
Material | Cellulose Nitrate |
Product Code | Brand | Diameter (Metric) Membrane | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Membrane | Quantity | Price | Quantity & Availability | ||||
10673321
|
Sartorius™
11406--50----ACN |
50 mm | 100 Pk. |
144.00€
Pack of 100 |
In Stock
Log in to see stock. |
||||
11790786
|
Sartorius™
11406--47----SCM |
47 mm | 300 Pk. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
- Product Type: Membrane Filter
- Material: Cellulose Nitrate
- Membrane Color/Grid Color: White/Black
- Pore Size: 0.45 μm
- Sterility: Sterile
- For Use With (Application): Bioburden/Microbial Enumeration, Food and Beverage, Environmental/Water Testing
Specifications
White | |
Membrane Filter | |
Cellulose Nitrate | |
0.45 μm | |
Bioburden/Microbial Enumeration, Food and Beverage, Environmental/Water Testing | |
Membrane Filter |
Single-use | |
Disc | |
Cellulose Nitrate | |
Sterile | |
Black |