Learn More
Bola X10 X10 Filtering Membranes 47mm 0.05um thickness(VE=10 Stck.)
Brand:  Bola N 1690-48
X10 X10 Filtering Membranes 47mm 0.05um thickness
Product Code. 10043501
  • ONLINE EXCLUSIVE PRICE
    67.10€
    54.37€ / Pack of 10
    Save 12.73€
    18% Off
Estimated Shipment: 18-01-2027