Learn More
KGW Isotherm™ Dewar Cylindrical Insulated Flasks
Product Code. 10433501
Change view
Click to view available options
Capacity (Metric):
300 mL
1 L
1500 mL
2 L
Includes:
No side grip
With side grip
Height (Metric) Inner:
190mm
235mm
240mm
245mm
290mm
340mm
Diameter (Metric) Inner:
47 mm
77 mm
90 mm
100 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes Height (Metric) Inner Diameter (Metric) Inner Unit Size
10433501 1 L No side grip 235mm 77 mm Each
10067890 300 mL No side grip 190mm 47 mm Each
10790232 1500 mL No side grip 240mm 100 mm Each
10558672 1500 mL With side grip 245mm 90 mm Each
10109560 1500 mL No side grip 245mm 90 mm Each
10740412 2 L No side grip 290mm 100 mm Each
10169600 2 L No side grip 340mm 90 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10433501 Supplier KGW Isotherm™ Supplier No. 10211
  • ONLINE EXCLUSIVE PRICE
    248.00€
    173.60€ / Pack of 1
    Save 74.40€
    30% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
For the storage and insulation of temperature-sensitive goods such as dry ice, liquid nitrogen and other coolants. KGW Isotherm™ Dewar Cylindrical Insulated Flasks feature a robust design made from borosilicate glass 3.3.
  • Easy handling
  • Cooling of small samples
  • Fully silvered as standard
  • Flexible applications in laboratory
  • Available with or without side grip
Specifications
Color Blue
Diameter (Metric) Outer 95mm
For Use With (Application) Cooling of small samples, storage of liquid nitrogen, dry ice, other coolants and as insulating flask
Height (Metric) Inner 235mm
Height (Metric) 290mm
Includes No side grip
Capacity (Metric) 1 L
Diameter (Metric) Inner 77 mm
Material Borosilicate Glass