Learn More
1-Heptyne, 99%
CAS: 628-71-7 | C7H12 | 96.173 g/mol
27.20€ - 199.00€
Chemical Identifiers
| CAS | 628-71-7 |
|---|---|
| Molecular Formula | C7H12 |
| Molecular Weight (g/mol) | 96.173 |
| MDL Number | MFCD00009529 |
| InChI Key | YVXHZKKCZYLQOP-UHFFFAOYSA-N |
| Synonym | 1-heptyne, amylacetylene, heptyne, unii-z3nf9ssh6k, z3nf9ssh6k, amyl acetylene, n-pentylacetylene, pubchem13058, acmc-1aw2o |
| PubChem CID | 12350 |
| IUPAC Name | hept-1-yne |
| SMILES | CCCCCC#C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11474170
|
Thermo Scientific Alfa Aesar
A11130.06 |
5 g |
27.20€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11489023
|
Thermo Scientific Alfa Aesar
A11130.14 |
25 g |
72.70€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11499023
|
Thermo Scientific Alfa Aesar
A11130.22 |
100 g |
199.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.
Solubility
Soluble in organic solvents. Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| C7H12 | |
| MFCD00009529 | |
| 1-heptyne, amylacetylene, heptyne, unii-z3nf9ssh6k, z3nf9ssh6k, amyl acetylene, n-pentylacetylene, pubchem13058, acmc-1aw2o | |
| hept-1-yne |
Specifications
| 628-71-7 | |
| 98°C to 100°C | |
| Unpleasant | |
| MFCD00009529 | |
| 1733518 | |
| YVXHZKKCZYLQOP-UHFFFAOYSA-N | |
| hept-1-yne | |
| 12350 | |
| 99% |
| -81°C | |
| −10°C (14°F) | |
| C7H12 | |
| UN3295 | |
| 1-heptyne, amylacetylene, heptyne, unii-z3nf9ssh6k, z3nf9ssh6k, amyl acetylene, n-pentylacetylene, pubchem13058, acmc-1aw2o | |
| CCCCCC#C | |
| 96.173 | |
| 96.17 | |
| 1-Heptyne |
Safety and Handling
GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
GHS P Statement
P210-P243-P301+P330+P331-P315-P403+P233
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.
Store in a well-ventilated place.
Keep container tightly closed.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
missing translation for 'tsca' : TSCA