Thermo Scientific Compact Digital Rocker
Ensure gentle sample agitation with our compact, adjustable angle, digital display unit.
Features:
- PID control ensures consistent and smooth rocking motion
- Digital speed control: 5 to 100rpm
- Easy-to-adjust tilt angle: 7° to 13°
- Compact design is ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C)
- Timer range: continuous or 1 minute to 99 hours, 59 minutes
- Includes dimpled mat (tray, silicone pad, two securing bands)
Applications
Designed for time-dependent and low-dependent sensitive applications such as ELISA, protein synthesis, staining/de-staining gels, and antibody staining.
Thermo Scientific Vari-Mix and Speci-Mix Test Tube Rockers
Offer a choice of fixed - or adjustable-angle models.
Features:
- All models hold 10 to 30mm diameter tubes and are ideal for blood collection tubes; Choose from fixed or adjustable speeds
- Reversible pad provides one side (with lip to prevent sliding) for capped tubes 110mm (4.33in.) long; other side for longer tubes
- Autoclavable white silicone rubber pad enhances viewing
- Can be used in a noncondensing incubator or coldroom with ambient operating conditions of 4° to 40°C (39.2° to 104°F) and 20 to 80% RH noncondensing
Thermo Scientific MaxQ 8000 Shakers
This video showcases the Thermo Scientific MaxQ 8000 Shakers.
Ease-Of-Use
Video Highlights of the Thermo Scientific MaxQ 8000 Shakers ease-of-use.
Easy Cleaning and Maintenance
Thermo Scientific MaxQ 8000 Easy Cleaning and Maintenance Video.
Ergonomic
Thermo Scientific MaxQ 8000 shakers - features an ergonomic fold down door and handle.
Flexible
The Thermo Scientific Max Q 8000 shaker plantforms featuer exceptional flexability.
HEPA Protection
Demonstration of the easily accessible HEPA Filter.
Intuitive Controls
MaxQ 8000 easy to program intuitive controls.
Sample Visibility
Large window and internal light allows for sample viewing.
Service Made Easy
Electronic Components and HEPA filter are accessed from the front of the shaker.
Thermo Scientific CO2 Resistant Digital Compact Bottle/Tube Roller
Maximize valuable bench space with our compact, adjustable-speed unit, capable of rolling multiple tube and bottle sizes.
Features
- Adjustable speed accommodates a variety of applications
- Digital speed control from 1 -80 RPM
- Small footprint ideal for use in a CO2 incubator
- Anti-corrosive and wear-resistant construction enables use in humid and CO2 environments
- Adjustable rollers accommodate a variety of tube/bottle sizes
- Stacking kit allows you to stack units two high, saving valuable bench space
- Optional remote control facilitates operating unit while in incubator and add timer function
- Includes pack of O-rings for tubes. O-rings ensure consistent smooth rolling when using tubes
Applications
Cell Culture | Hybridization Assays | Liquid Mixing
Thermo Scientific Digital Tube Revolver
Increase your lab's efficiency with our variable-speed tube revolver. Its small footprint, flexible design and simple display help increase flexibility and effectiveness.
Features
- Interchangeable rotisseries accommodate different tube sizes for maximum flexibility. Each paddle can be adjusted from 0-90 degrees
- Achieve more effective mixing with "oscillating" mode. Oscillation will occur at every 40 degrees in the rotation cycle
- Small footprint
- Plug-and-play simplicity
- Maintenance-free motor, quiet operation
Standard Unit Includes:
- 90-691-36 — 10mL/15mL paddle, Quantity 2
- 90-691-37 — 5mL/7mL paddle, Quantity 2
- 90-691-38 — 1.5mL/2mL paddle, Quantity 2
- 90-691-40 — 50mL paddle, Quantity 1
Applications
Molecular Biology | Clinical Biochemistry
Thermo Scientific Compact Digital Waving Rotator
Achieve maximum sample mixing efficiency with the adjustable orbital waving digital-display rotator.
Features:
- PID control ensures consistent and smooth rocking motion
- Digital Speed Control: 8 to100rpm
- Easy to adjust tilt angle: 0° to 13°
- Compact design ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C)
- Timer range: continuous or 1 minute to 99 hours 59 minutes
- Includes dimpled mat (tray, silicone pad, two securing bands)
- When no angle is used, the Waving Rotator can be used as a slow speed orbital shaker
Applications
Designed for time-dependent and low-dependent sensitive applications such as ELISA, protein synthesis, staining/de-staining gels and antibody staining.
Thermo Scientific Cel-Gro Tissue Culture Rotator
Available in a single or dual drum configuration.
Features
- Single-drum as an adjustable rotating angle and speed, while the dual-drum has a fixed rotating angle and speed
- Rotator operates in standard incubators for cultivation under controlled temperature conditions
- Powder-coated stainless-steel outer body construction is durable and corrosion-resistant
- Drums can be lifted off the rotor without interrupting rotator operation
- Adjusting screw on the base varies elevation angle
- Adjustable 5° rotating angle above horizontal to 90°
- Adjustable speed control from 12 to 70rpm
- Drum required with purchase, sizes vary based on tube size requirements:
- 142 x 17mm tube diameter
- 76 x 26mm tube diameter
- 60 x 30mm tube diameter
Thermo Scientific LP Vortex Mixer
Choose to continuously mix or pulse samples with our low-profile vortex mixer.
Features:
- Low-profile design with small footprint
- Run in continuous or pulsing mode
- Speed control: 0 to 300rpm
- Automatic overload shut off
- Wide variety of optional accessories to accommodate different vessel types
Applications
Any application that requires intense vortex mixing in a tube, plate or glass vessel.
Thermo Scientific MaxQ Orbital Shakers: A Shaker for Every Lab
You have important discoveries to make. Thermo Scientific has your shaker. Designs large or small, open air, incubated, or refrigerated, reciprocating or orbital, benchtop or floor models — Thermo Scientific has shakers to fit your lab and your samples, your ideas.
Thermo Scientific offers platforms and accessories for almost any application and vessel type to give you maximum flexibility. Backlit, intuitive displays make it easy to see all parameters at once. If you want to shake things up but stay in control, choose Thermo Scientific MaxQ Shakers.
Select Your Platform
Universal Platforms
Universal platforms allow you to maximize flexibility in the lab by supporting a mixture of different size clamps from standard Erlenmeyer flask clamps to microplate and test tube clamps. Universal platforms are ideal for shared equipment on which various applications are being performed.
In addition, the universal platform allows the shaker to change with the needs of the lab.
Dedicated Platforms
Dedicated platforms are designed to maximize capacity for a single vessel size only. Select the pre-configured dedicated platform based upon the vessel you need to shake.
Dual-Tiered Platforms
Dual-tiered platforms are designed to double your capacity in the footprint of one shaker. Dual tiered platforms are only available for the Thermo Scientific MaxQ 2000, MaxQ 3000 and MaxQ 6000. The platforms are a universal format to allow for a wide range of vessels.
Browse dual-tiered platforms for Thermo Scientific MaxQ 2000, 2506 and 2508
Browse dual-tiered platforms for Thermo Scientific MaxQ 2000, 2506, 2508 and 6000
Browse dual-tiered platforms for Thermo Scientific MaxQ 3000
Select Your Clamps
Select the clamps you need to accommodate your vessels from the universal clamp selection. Clamps are available for Erlenmeyer flasks, Fernbach flasks, microplates and test tube racks. In addition, adhesive flask mats and adjustable vessel dedicated platforms are available to accommodate non-traditional vessels you wish to shake.
Browse Thermo Scientific shaker clamps
MoreEven More Shaker Options
Thermo Scientific Compact Digital Microplate Shaker
Harness the efficiency of our microplate shaker, capable of shaking up to four microplates in one run.
Features:
- PID control ensures consistent shaking results and precise temperature control
- Digital display for easy viewing
- Compact design is ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C)
- Timer range: continuous or 1 minute to 99 hours, 59 minutes
- Automatic overload shutdown
- 4.4kg / 9.7lbs
Applications
Any application requiring precise and accurate shaking of microplates: immunoassay, screening applications, magnetic bead applications
Thermo Scientific CO2 Resistant Shakers
Reliable around-the-clock operation.
Features:
- Orbital shaking through magnetic power eliminates the need to replace belts
- Small footprint ideal for use in a CO2 incubator
- Specially treated mechanical components protect up to 20% CO2 and 95% humidity conditions
- Convenient external control box with LED display allows for easy adjustments without opening the chamber door
Thermo Scientific Compact Digital Orbital Shaker
Realize maximum flexibility with the easy-to-use, digitally controlled rotator that provides smooth, quiet orbital shaking of different tube/vessel types.
Features:
- PID control ensures a consistent and smooth motion
- Digital speed control: 50 to 300rpm
- Timer range: continuous or 1 minute to 99 hours 59 minutes
- Wide variety of accessories to customize unit for exact laboratory needs
- Compact design ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C)
- Must purchase a platform with unit, universal attachment requires clamps
Applications
Bacteria and Fungi Culturing | Hybridization assays | Gel Staining/De-Staining
Thermo Scientific Digital Heating Shaking Dry Bath
Increase lab versatility with the Digital Shaking Dry Bath, which can be used as a Dry Bath, Shaker or both.
Features:
- PID control ensures consistent shaking results and precise temperature control
- Speed range from 150 to 1500rpm (block dependent)
- Program storage
- 10 programs, up to 10 steps per program
- Interval shaking function allows shaking and non-shaking in same program
- Two types of timing modes
- Start only after temperature reaches desired set point
- Start immediately after the timer is set
- Easily changed blocks in multiple sizes allow for maximum flexibility
- Note: A block must be purchased with the unit
Applications
DNA Extractions | Enzymatic Reactions
Thermo Scientific MaxQ Shakers Brochure
The Thermo Scientific MaxQ Shaker product selector guide will help you build the optimal shaker for your lab and application.
Thermo Scientific Rockers, Rotators and Shakers Brochure
The Thermo Scientific Rockers, Rotators and Shakers brochure includes a list of products along with ordering information and product specifications.