CAS RN 99277-71-1
CAS RN 99277-71-1
IUPAC Name:
4-bromo-2-nitrobenzoate
Synonyms:
4-bromo-2-nitrobenzoate
Molecular Weight (g/mol):
245.01
Molecular Formula:
C7H3BrNO4
InChi Key:
ZIRHHEZLJGORGU-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(Br)C=C1[N+]([O-])=O
1
–
1
of
1
results
1
4-Bromo-2-nitrobenzoic acid, 97%
CAS: 99277-71-1 Molecular Formula: C7H3BrNO4 Molecular Weight (g/mol): 245.01 MDL Number: MFCD01013599 InChI Key: ZIRHHEZLJGORGU-UHFFFAOYSA-M Synonym: 4-bromo-2-nitrobenzoic acid,4-bromo-2-nitro-benzoic acid,2-nitro-4-bromobenzoic acid,5-bromo-2-carboxynitrobenzene,benzoic acid, 4-bromo-2-nitro,4-bromo-2-nitrobenzoicacid,pubchem2055,4-bromo-2-nitrobenzoic,acmc-209sc4,bromonitrobenzoicacid 42- PubChem CID: 3774467 IUPAC Name: 4-bromo-2-nitrobenzoate SMILES: [O-]C(=O)C1=CC=C(Br)C=C1[N+]([O-])=O