accessibility menu, dialog, popup

UserName

H3COCl

CAS RN 99-02-5

IUPAC Name: 1-(3-chlorophenyl)ethan-1-one
Synonyms: M-chloroacetophenone chloroacetophenone (mixture of isomers)
Molecular Weight (g/mol): 154.59
Molecular Formula: C8H7ClO
InChi Key: UUWJBXKHMMQDED-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(Cl)=C1
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)