accessibility menu, dialog, popup

UserName

CAS RN 96693-05-9

H3COBr

CAS RN 96693-05-9

IUPAC Name: 1-bromo-4-(octyloxy)benzene
Synonyms: 1-bromo-4-(octyloxy)benzene
Molecular Weight (g/mol): 285.23
Molecular Formula: C14H21BrO
InChi Key: UVBFFPZGOOKWNR-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=C(Br)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results