accessibility menu, dialog, popup

UserName

NNBrH2NBr

CAS RN 957230-70-5

IUPAC Name: 3,6-dibromopyrazin-2-amine
Synonyms: 3,6-dibromopyrazin-2-amine
Molecular Weight (g/mol): 252.9
Molecular Formula: C4H3Br2N3
InChi Key: ZEDFFSRZLCREEN-UHFFFAOYSA-N
SMILES: NC1=NC(Br)=CN=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results