accessibility menu, dialog, popup

UserName

CAS RN 956-04-7

ClO(E)

CAS RN 956-04-7

IUPAC Name: (2E)-3-(4-chlorophenyl)-1-phenylprop-2-en-1-one
Synonyms: 4-chlorochalcone C15H11ClO
Molecular Weight (g/mol): 242.7
Molecular Formula: C15H11ClO
InChi Key: ABGIIXRNMHUKII-DHZHZOJOSA-N
SMILES: ClC1=CC=C(\C=C\C(=O)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results