CAS RN 95453-58-0
IUPAC Name:
2-chloro-1,3-thiazole-5-carbaldehyde
Synonyms:
2-chloro-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol):
147.58
Molecular Formula:
C4H2ClNOS
InChi Key:
PKCBQQXHFIDIIG-UHFFFAOYSA-N
SMILES:
ClC1=NC=C(S1)C=O
1
–
1
of
1
results
1
2-Chlorothiazole-5-carboxaldehyde, 97%, Thermo Scientific™
CAS: 95453-58-0 Molecular Formula: C4H2ClNOS Molecular Weight (g/mol): 147.58 MDL Number: MFCD01568717 InChI Key: PKCBQQXHFIDIIG-UHFFFAOYSA-N Synonym: 2-chlorothiazole-5-carbaldehyde,2-chloro-1,3-thiazole-5-carboxaldehyde,2-chlorothiazole-5-carboxaldehyde,5-thiazolecarboxaldehyde, 2-chloro,2-chloro-5-thiazolecarboxaldehyde,2-chloro-thiazole-5-carbaldehyde,2-chloro-5-formylthiazole,2-chloro-5-formyl-1,3-thiazole,2-chloro-1,3-thiazol-5-yl formaldehyde,5-thiazolecarboxaldehyde, 2-chloro-9ci PubChem CID: 1479765 IUPAC Name: 2-chloro-1,3-thiazole-5-carbaldehyde SMILES: ClC1=NC=C(S1)C=O