accessibility menu, dialog, popup

UserName

NH2(S)NOOOHO

CAS RN 949-99-5

IUPAC Name: (2S)-2-amino-3-(4-nitrophenyl)propanoic acid
Synonyms: phenylalanine, 4-nitro-
Molecular Weight (g/mol): 210.19
Molecular Formula: C9H10N2O4
InChi Key: GTVVZTAFGPQSPC-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)