accessibility menu, dialog, popup

UserName

CAS RN 947536-03-0

CH3ONONNH3CNNOOFFF

CAS RN 947536-03-0

IUPAC Name: N-{1-[3-(4-ethoxyphenyl)-4-oxo-3H,4H-pyrido[2,3-d]pyrimidin-2-yl]ethyl}-N-[(pyridin-3-yl)methyl]-2-[4-(trifluoromethoxy)phenyl]acetamide
Synonyms: N-{1-[3-(4-ethoxyphenyl)-4-oxopyrido[2,3-d]pyrimidin-2-yl]ethyl}-N-(pyridin-3-ylmethyl)-2-[4-(trifluoromethoxy)phenyl]acetamide
Molecular Weight (g/mol): 603.6
Molecular Formula: C32H28F3N5O4
InChi Key: WQTKNBPCJKRYPA-UHFFFAOYNA-N
SMILES: CCOC1=CC=C(C=C1)N1C(=O)C2=CC=CN=C2N=C1C(C)N(CC1=CN=CC=C1)C(=O)CC1=CC=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results