accessibility menu, dialog, popup

UserName

CAS RN 94741-69-2

NH2NClNN

CAS RN 94741-69-2

IUPAC Name: 4-amino-2-chloropyrimidine-5-carbonitrile
Synonyms: 4-amino-2-chloropyrimidine-5-carbonitrile
Molecular Weight (g/mol): 154.56
Molecular Formula: C5H3ClN4
InChi Key: WDHFCSOENXEMRC-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=NC=C1C#N
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results