accessibility menu, dialog, popup

UserName

CAS RN 94651-33-9

FFFOO

CAS RN 94651-33-9

IUPAC Name: 2-(trifluoromethoxy)benzaldehyde
Synonyms: 2-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 190.12
Molecular Formula: C8H5F3O2
InChi Key: CPHXLFKIUVVIOQ-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1C=O
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)