accessibility menu, dialog, popup

UserName

CAS RN 946409-19-4

CH3NNONH2

CAS RN 946409-19-4

IUPAC Name: 1-[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methanamine
Synonyms: 1-[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methanamine
Molecular Weight (g/mol): 189.22
Molecular Formula: C10H11N3O
InChi Key: SBNUQIHHPTZLSV-UHFFFAOYSA-N
SMILES: CC1=NN=C(O1)C1=CC=C(CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results