CAS RN 943962-47-8
CAS RN 943962-47-8
IUPAC Name:
3,5-dichloro-2-hydroxy-N-{4-methoxy-[1,1'-biphenyl]-3-yl}benzene-1-sulfonamide
Synonyms:
3,5-dichloro-2-hydroxy-N-{4-methoxy-[1,1'-biphenyl]-3-yl}benzenesulfonamide
Molecular Weight (g/mol):
424.29
Molecular Formula:
C19H15Cl2NO4S
InChi Key:
SIIPNDKXZOTLEA-UHFFFAOYSA-N
SMILES:
COC1=C(NS(=O)(=O)C2=CC(Cl)=CC(Cl)=C2O)C=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ BMS 303141
CAS: 943962-47-8 Molecular Formula: C19H15Cl2NO4S