accessibility menu, dialog, popup

UserName

NNO

CAS RN 94378-00-4

IUPAC Name: 2-(furan-2-yl)-1,3-diphenylimidazolidine
Synonyms: 2-(furan-2-yl)-1,3-diphenylimidazolidine
Molecular Weight (g/mol): 290.37
Molecular Formula: C19H18N2O
InChi Key: GGKPIWBVFHMQMP-UHFFFAOYSA-N
SMILES: C1CN(C(N1C1=CC=CC=C1)C1=CC=CO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results