accessibility menu, dialog, popup

UserName

CAS RN 942-92-7

IUPAC Name: 1-phenylhexan-1-one
Synonyms: 1-hexanone, 1-phenyl-
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: MAHPVQDVMLWUAG-UHFFFAOYSA-N
SMILES: CCCCCC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)