CAS RN 936-08-3
CAS RN 936-08-3
IUPAC Name:
2-bromo-4-chlorobenzoic acid
Synonyms:
2-bromo-4-chlorobenzoic acid
Molecular Weight (g/mol):
235.46
Molecular Formula:
C7H4BrClO2
InChi Key:
USMQLFCVCDEXAK-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(Br)C=C(Cl)C=C1
1
–
1
of
1
results
1
2-Bromo-4-chlorobenzoic acid, 97%
CAS: 936-08-3 Molecular Formula: C7H4BrClO2 Molecular Weight (g/mol): 235.461 MDL Number: MFCD00672930 InChI Key: USMQLFCVCDEXAK-UHFFFAOYSA-N Synonym: 2-bromo-4-chloro-benzoic acid,benzoic acid, 2-bromo-4-chloro,2-bromo-4-chlorobenzoicacid,pubchem4028,acmc-209rmm,p-chloro bromo benzoic acid,ksc486m1d,benzoicacid, 2-bromo-4-chloro,2-bromo-4-chlorobenzoic acid PubChem CID: 33126 IUPAC Name: 2-bromo-4-chlorobenzoic acid SMILES: C1=CC(=C(C=C1Cl)Br)C(=O)O