accessibility menu, dialog, popup

UserName

CAS RN 93560-55-5

H3CONI

CAS RN 93560-55-5

IUPAC Name: 2-iodo-3-methoxypyridine
Synonyms: 2-iodo-3-methoxypyridine
Molecular Weight (g/mol): 235.02
Molecular Formula: C6H6INO
InChi Key: NJFRZBAZMPWJKQ-UHFFFAOYSA-N
SMILES: COC1=C(I)N=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results