CAS RN 935-13-7
IUPAC Name:
3-(furan-2-yl)propanoate
Synonyms:
3-(furan-2-yl)propanoate
Molecular Weight (g/mol):
139.13
Molecular Formula:
C7H7O3
InChi Key:
XLTJXJJMUFDQEZ-UHFFFAOYSA-M
SMILES:
[O-]C(=O)CCC1=CC=CO1
1
–
2
of
2
results
2
3-(2-Furyl)propanoic acid, 98%
CAS: 935-13-7 Molecular Formula: C7H8O3 Molecular Weight (g/mol): 140.14 InChI Key: XLTJXJJMUFDQEZ-UHFFFAOYSA-N Synonym: 3-furan-2-yl propanoic acid,3-2-furyl propanoic acid,2-furanpropanoic acid,2-furanpropionic acid,3-2-furyl propionic acid,3-furan-2-yl-propionic acid,furan-2-propionic acid,3-2-furyl propoinic acid,furfuryl-essigsaure,furfurylacetic acid PubChem CID: 70286 IUPAC Name: 3-(furan-2-yl)propanoic acid SMILES: C1=COC(=C1)CCC(=O)O